2-Amino-5-phenylpyridine ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Phenylpyridin-2-amine, 5-Phenyl-2-pyridylamine

007056-1G 007056-5G
101791-918EA 46.15 USD
101791-918 101791-920
2-Amino-5-phenylpyridine ≥97%
2-Amino-5-phenylpyridine

Hazard code(s): Irritant

Formula: C₁₁H₁₀N₂
MW: 170.21 g/mol
Melting Pt: 132…133 °C
MDL Number: MFCD01692452
CAS Number: 33421-40-8

Order Now

Specification Test Results

Melting point 132-133°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR