3-Aminobenzophenone ≥98%

Supplier: Matrix Scientific

Synonyms: m-Aminobenzophenone

007053-5G 007053-25G
101791-910EA 29.23 USD
101791-910 101791-912
3-Aminobenzophenone ≥98%
3-Aminobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₁₁NO
MW: 197.24 g/mol
Melting Pt: 85…87 °C
MDL Number: MFCD00017103
CAS Number: 2835-78-1
EINECS: 220-614-4

Order Now

Specification Test Results

Melting point 85-87°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR