4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol ≥97%

Supplier: Matrix Scientific

007046-5G 007046-25G
101791-890EA 453.85 USD
101791-890 101791-892
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol ≥97%
4,4,5,5,6,6,7,7,7-Nonafluoro-1-heptanol

Hazard code(s): Irritant

Formula: C₇H₇F₉O
Boiling Pt: 46…48 °C (1mm Hg)
Density: 1.528 g/cm³
MDL Number: MFCD02183552
CAS Number: 83310-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR