Methyl-3,5-dichlorobenzoate

Supplier: Matrix Scientific

Synonyms: Methyl 3,5-dichlorobenzoate

007030-1G 007030-5G
101791-870EA 21.54 USD
101791-870 101791-872
Methyl-3,5-dichlorobenzoate
Methyl-3,5-dichlorobenzoate
Formula: C₈H₆Cl₂O₂
MW: 205.04 g/mol
MDL Number: MFCD00032710
CAS Number: 2905-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR