2,4-Difluorobenzhydrazide

Supplier: Matrix Scientific

Synonyms: 2,4-Difluorobenzoic acid hydrazide, 2,4-Difluorobenzohydrazide, 2,4-Difluorobenzoic hydrazide, 2,4-Difluorobenzene-1-carbohydrazide

007015-1G 007015-5G
101791-830EA 26.15 USD
101791-830 101791-832
2,4-Difluorobenzhydrazide
2,4-Difluorobenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₆F₂N₂O
MW: 172.13 g/mol
Melting Pt: 141…142 °C
MDL Number: MFCD00173685
CAS Number: 118737-62-5

Order Now

Specification Test Results

Melting point 141-142°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR