3,5-Dichlorobenzhydrazide

Supplier: Matrix Scientific

Synonyms: 3,5-Dichlorobenzoic hydrazide

007010-1G 007010-5G
101791-820EA 104.62 USD
101791-820 101791-822
3,5-Dichlorobenzhydrazide
3,5-Dichlorobenzhydrazide

Hazard code(s): Irritant

Formula: C₇H₆Cl₂N₂O
MW: 205.04 g/mol
Melting Pt: 205…207 °C
MDL Number: MFCD00833411
CAS Number: 62899-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR