4-Chlorobenzoylacetonitrile
Supplier: Matrix Scientific
007002-5G
007002-25G
101791-794EA
36.92
USD
101791-794
101791-796
4-Chlorobenzoylacetonitrile
4-Chlorobenzoylacetonitrile
Hazard code(s): Irritant, Irritant-Harmful
Formula:
C₉H₆ClNO Melting Pt: 129…133 °C |
MDL Number:
MFCD00051625 CAS Number: 4640-66-8 |
Specification Test Results
Melting point | 129-133°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...