6-Nitroindoline ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 2,3-Dihydro-6-nitro-(1H)-indole

006994-1G 006994-5G
101791-772EA 21.54 USD
101791-772 101791-774
6-Nitroindoline ≥97%
6-Nitroindoline

Hazard code(s): Irritant

Formula: C₈H₈N₂O₂
MW: 164.16 g/mol
Melting Pt: 65…67 °C
MDL Number: MFCD00005710
CAS Number: 19727-83-4
EINECS: 243-257-6

Order Now

Specification Test Results

Melting point 65-67°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR