6-Methoxyanthranilic acid ≥97%

Supplier: Matrix Scientific

006988-5G 006988-25G
101791-750EA 44.62 USD
101791-750 101791-752
6-Methoxyanthranilic acid ≥97%
6-Methoxyanthranilic acid

Hazard code(s): Irritant

Formula: C₈H₉NO₃
MW: 167.16 g/mol
Melting Pt: >85 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01941328
CAS Number: 53600-33-2

Order Now

Specification Test Results

Melting point >85°C(dec)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR