5-Amino-2-(trifluoromethyl)pyridine ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Amino-6-(trifluoromethyl)pyridine, 2-(Trifluoromethyl)pyridin-5-amine, 2-(Trifluoromethyl)-5-pyridylamine

006911-5G 006911-25G
101791-542EA 64.62 USD
101791-542 101791-544
5-Amino-2-(trifluoromethyl)pyridine ≥98%
5-Amino-2-(trifluoromethyl)pyridine

Hazard code(s): Irritant

Formula: C₆H₅F₃N₂
MW: 162.11 g/mol
Boiling Pt: 92…94 °C (0.01mm)
Melting Pt: 39…40 °C
MDL Number: MFCD01862656
CAS Number: 106877-33-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Boiling point 92-94°C/0.0
Melting point 39-40°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR