7-Methylindole ≥98% for synthesis

Supplier: Matrix Scientific
Warning

006871-5G 006871-25G 006871-100G
101791-406EA 113.85 USD
101791-406 101791-408 101791-410
7-Methylindole ≥98% for synthesis
7-Methylindole

Hazard code(s): Irritant

Formula: C₉H₉N
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.0202 g/cm³ (99.6 °C)
MDL Number: MFCD00005684
CAS Number: 933-67-5
EINECS: 213-270-1

Order Now

Specification Test Results

Boiling point 85-89°C/0.2
Melting point 79-81°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR