2,3-Dimethyl-7-nitroindole

Supplier: Matrix Scientific

Synonyms: 2,3-Dimethyl-7-nitro-1H-indole

006869-1G 006869-5G
101791-398EA 53.85 USD
101791-398 101791-400
2,3-Dimethyl-7-nitroindole
2,3-Dimethyl-7-nitroindole

Hazard code(s): Irritant

Formula: C₁₀H₁₀N₂O₂
MW: 190.2 g/mol
Melting Pt: 164…165 °C
MDL Number: MFCD00022710
CAS Number: 41018-86-4

Order Now

Specification Test Results

Melting point 164-165°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR