7-Methoxyindole ≥97%

Supplier: Matrix Scientific
Warning

Synonyms: 7-Methoxy-1H-indole, Indol-7-yl methyl ether

006853-1G 006853-5G
101791-336EA 58.46 USD
101791-336 101791-338
7-Methoxyindole ≥97%
7-Methoxyindole

Hazard code(s): Light Sensitive, Refrigerate

Formula: C₉H₉NO
MW: 147.18 g/mol
Boiling Pt: 106 °C (0.4mm)
Density: 1.126 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD00047203
CAS Number: 3189-22-8
EINECS: 221-690-1

Order Now

Specification Test Results

Boiling point 106°C/0.4mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR