4-Bromoindole ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-1H-indole

006845-5G 006845-25G 006845-100G
101791-302EA 87.69 USD
101791-302 101791-304 102832-780
4-Bromoindole ≥95%
4-Bromoindole

Hazard code(s): Irritant, Refrigerate

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 88…90 °C (0.1mm)
Melting Pt: 17 °C
Density: 1.563 g/cm³
MDL Number: MFCD00671502
CAS Number: 52488-36-5

Order Now

Specification Test Results

Boiling point 88-90°C/0.1
Melting point 17°C
Density 1.563
ND20 1.655

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR