6-Chloroindole ≥97%

Supplier: Matrix Scientific

Synonyms: 6-Chloro-1H-indole

006844-1G 006844-5G
101791-296EA 15.38 USD
101791-296 101791-298
6-Chloroindole ≥97%
6-Chloroindole

Hazard code(s): Irritant

Formula: C₈H₆ClN
MW: 151.6 g/mol
Melting Pt: 86…88 °C
MDL Number: MFCD00005681
CAS Number: 17422-33-2
EINECS: 241-449-4

Order Now

Specification Test Results

Melting point 86-88°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR