2-Chloro-4,6-dimethylpyrimidine ≥97%

Supplier: Matrix Scientific

006836-5G 006836-25G
101791-276EA 21.54 USD
101791-276 101791-278
2-Chloro-4,6-dimethylpyrimidine ≥97%
2-Chloro-4,6-dimethylpyrimidine

Hazard code(s): Irritant

Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Melting Pt: 34…36 °C
MDL Number: MFCD00023199
CAS Number: 4472-44-0

Order Now

Specification Test Results

Melting point 34-36°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR