Indole-6-carboxylic acid ≥98%

Supplier: Matrix Scientific

Synonyms: 1H-Indole-6-carboxylic acid

006806-5G 006806-25G 006806-100G
101791-164EA 21.54 USD
101791-164 101791-166 101791-168
Indole-6-carboxylic acid ≥98%
Indole-6-carboxylic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₉H₇NO₂
MW: 161.16 g/mol
Melting Pt: 254 - 257 °C
MDL Number: MFCD00210441
CAS Number: 1670-82-2

Order Now

Specification Test Results

Melting point 256-259°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR