2-Naphthyl boronic acid ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
2-Naphthaleneboronic acid, Naphthalene-2-boronic acid, 2-Naphthyl boric acid
006788-5G
006788-25G
006788-100G
101791-114EA
15.38
USD
101791-114
101791-116
101791-118
2-Naphthyl boronic acid ≥97%
2-Naphthyl boronic acid
Hazard code(s): Irritant, Store in Freezer
Formula:
C₁₀H₉BO₂ MW: 171.99 g/mol |
MDL Number:
MFCD00236051 CAS Number: 32316-92-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...