4-Ethylphenylboronic acid ≥97% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: (4-Ethylphenyl)boronic acid, (4-Ethylphenyl)metaboric acid, 4-Ethylbenzeneboronic acid, Dihydroxy(4-ethylphenyl)borane

006779-5G 006779-25G 006779-100G
101791-094EA 15.38 USD
101791-094 101791-096 101791-098
4-Ethylphenylboronic acid ≥97% for synthesis
4-Ethylphenylboronic acid

Hazard code(s): Irritant, Store in Freezer

Formula: C₈H₁₁BO₂
MW: 149.98
Density: 1.125 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00859377
CAS Number: 63139-21-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR