4-Ethoxyphenylboronic acid ≥97%

Supplier: Matrix Scientific

006778-1G 006778-5G
101791-090EA 15.38 USD
101791-090 101791-092
4-Ethoxyphenylboronic acid ≥97%
4-Ethoxyphenylboronic acid

Hazard code(s): Irritant, Refrigerate

Formula: C₈H₁₁BO₃
MDL Number: MFCD00674028
CAS Number: 22237-13-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR