5-Bromo-2-methoxypyridine ≥97%

Supplier: Matrix Scientific
Warning

006766-25G 006766-100G
101791-050EA 29.23 USD
101791-050 101791-052
5-Bromo-2-methoxypyridine ≥97%
5-Bromo-2-methoxypyridine

Hazard code(s): Irritant

Formula: C₆H₆BrNO
MW: 188.02 g/mol
Boiling Pt: 80 °C (12mm)
Density: 1.453 g/cm³
MDL Number: MFCD01318952
CAS Number: 13472-85-0

Order Now

Specification Test Results

Boiling point 80°C/12mm
Density 1.453
ND20 1.555

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR