2-Bromo-N,N-dimethylaniline ≥97%

Supplier: Matrix Scientific

006765-5G 006765-25G
101791-046EA 60 USD
101791-046 101791-048
2-Bromo-N,N-dimethylaniline ≥97%
2-Bromo-N,N-dimethylaniline

Hazard code(s): Irritant

Formula: C₈H₁₀BrN
MW: 200.08 g/mol
Boiling Pt: 107 °C (14mm)
Melting Pt: 38 °C
Density: 1.388 g/cm³ (25 °C)
MDL Number: MFCD00013522
CAS Number: 698-00-0

Order Now

Specification Test Results

Boiling point 107°C/14mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR