5-Bromo-2-chloropyridine ≥98%

Supplier: Matrix Scientific
Warning

006764-25G 006764-100G
101791-042EA 29.23 USD
101791-042 101791-044
5-Bromo-2-chloropyridine ≥98%
5-Bromo-2-chloropyridine

Hazard code(s): Irritant, Irritant-Harmful

Formula: C₅H₃BrClN
MW: 192.44 g/mol
Boiling Pt: 205 °C (1013 hPa)
Melting Pt: 72…74 °C
MDL Number: MFCD01318951
CAS Number: 53939-30-3

Order Now

Specification Test Results

Melting point 72-74°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR