8-Aminoquinoline ≥97%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
Quinolin-8-ylamine
006752-5G
006752-25G
006752-100G
101791-012EA
21.54
USD
101791-012
101791-014
101791-016
8-Aminoquinoline ≥97%
8-Aminoquinoline
Hazard code(s): Irritant, Air Sensitive
Formula:
C₉H₈N₂ MW: 144.18 g/mol Boiling Pt: 290 °C (1013 hPa) Melting Pt: 64 °C Density: 1.337 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00006809 CAS Number: 578-66-5 EINECS: 209-427-9 |
Specification Test Results
Boiling point | 174°C/26mm |
Melting point | 65-68°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...