3-Methylbenzo[b]thiophene ≥96%

Supplier: Matrix Scientific

Synonyms: 3-Methylbenzothiophene

006734-5G 006734-25G
101790-986EA 63.08 USD
101790-986 101790-988
3-Methylbenzo[b]thiophene ≥96%
3-Methylbenzo[b]thiophene

Hazard code(s): Irritant

Formula: C₉H₈S
MW: 148.23 g/mol
Boiling Pt: 72…74 °C (2mm)
MDL Number: MFCD00040243
CAS Number: 1455-18-1
EINECS: 215-934-6

Order Now

Specification Test Results

Boiling point 72-74°C/2mm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR