4,5-Dibromo-o-xylene ≥98% for synthesis

Supplier: Matrix Scientific
Warning

Synonyms: 1,2-Dibromo-4,5-dimethylbenzene, 4,5-Dibromo-1,2-dimethylbenzene

006727-1G 006727-5G
101790-972EA 38.46 USD
101790-972 102550-878
4,5-Dibromo-o-xylene ≥98% for synthesis
4,5-Dibromo-o-xylene

Hazard code(s): Irritant

Formula: C₈H₈Br₂
Melting Pt: 85…87 °C
MDL Number: MFCD00082743
CAS Number: 24932-48-7

Order Now

Specification Test Results

Melting point 85-87°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR