2-Chloro-5-nitrobenzaldehyde ≥95%

Supplier: Matrix Scientific

006725-25G 006725-100G
101790-968EA 26.15 USD
101790-968 101790-970
2-Chloro-5-nitrobenzaldehyde ≥95%
2-Chloro-5-nitrobenzaldehyde

Hazard code(s): Irritant

Formula: C₇H₄ClNO₃
Melting Pt: 75 - 78 °C
Storage Temperature: Ambient
MDL Number: MFCD00007293
CAS Number: 6361-21-3
EINECS: 228-830-0

Order Now

Specification Test Results

Melting point 76-77°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR