2-Amino-5-bromo-3-nitropyridine ≥98%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
5-Bromo-3-nitropyridin-2-amine, 5-Bromo-3-nitro-2-pyridylamine
006718-5G
006718-25G
101790-952EA
21.54
USD
101790-952
101790-954
2-Amino-5-bromo-3-nitropyridine ≥98%
2-Amino-5-bromo-3-nitropyridine
Hazard code(s): Irritant
Formula:
C₅H₄BrN₃O₂ MW: 218.01 g/mol Melting Pt: 208…210 °C |
MDL Number:
MFCD00047441 CAS Number: 6945-68-2 |
Specification Test Results
Melting point | 208-210°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...