2-Acetyl-5-nitrobenzofuran ≥98%

Supplier: Matrix Scientific

006717-1G 006717-5G
101790-948EA 41.54 USD
101790-948 101790-950
2-Acetyl-5-nitrobenzofuran ≥98%
2-Acetyl-5-nitrobenzofuran

Hazard code(s): Irritant

Formula: C₁₀H₇NO₄
MW: 205.17 g/mol
Melting Pt: 175 °C
MDL Number: MFCD00051634
CAS Number: 23136-39-2

Order Now

Specification Test Results

Melting point 175°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR