4-Iodobenzophenone ≥98%

Supplier: Matrix Scientific

006692-1G 006692-5G
101790-932EA 63.08 USD
101790-932 101790-934
4-Iodobenzophenone ≥98%
4-Iodobenzophenone

Hazard code(s): Irritant

Formula: C₁₃H₉IO
MW: 308.12 g/mol
Melting Pt: 97…99 °C
MDL Number: MFCD00270109
CAS Number: 6136-66-9

Order Now

Specification Test Results

Melting point 97-99°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR