4-Iodobenzonitrile ≥98%

Supplier: Matrix Scientific

006690-25G 006690-100G
101790-924EA 53.85 USD
101790-924 101790-926
4-Iodobenzonitrile ≥98%
4-Iodobenzonitrile

Hazard code(s): Irritant

Formula: C₇H₄IN
Boiling Pt: 285 °C (1013 hPa)
Melting Pt: 126…128 °C
MDL Number: MFCD00051310
CAS Number: 3058-39-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 126-128°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR