3-Iodobenzonitrile ≥97%

Supplier: Matrix Scientific

006689-5G 006689-25G
101790-918EA 29.23 USD
101790-918 101790-920
3-Iodobenzonitrile ≥97%
3-Iodobenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₇H₄IN
Melting Pt: 35…37 °C
MDL Number: MFCD00079762
CAS Number: 69113-59-3
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Melting point 35-37°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR