2-Iodobenzonitrile ≥98%

Supplier: Matrix Scientific

006688-5G 006688-25G 006688-100G
101790-912EA 15.38 USD
101790-912 101790-914 101790-916
2-Iodobenzonitrile ≥98%
2-Iodobenzonitrile

Hazard code(s): Irritant-Harmful

Formula: C₇H₄IN
Melting Pt: 52…54 °C
MDL Number: MFCD00079761
CAS Number: 4387-36-4

Order Now

Specification Test Results

Melting point 52-54°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR