3-Bromostyrene ≥97% stabilized

Supplier: Matrix Scientific

Synonyms: 1-Bromo-3-vinylbenzene

006667-5G 006667-25G
101790-886EA 76.92 USD
101790-886 101790-888
3-Bromostyrene ≥97% stabilized
3-Bromostyrene

Hazard code(s): Irritant, Refrigerate

Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 74…75 °C (3mm)
Density: 1.4059 g/cm³ (20 °C)
MDL Number: MFCD00000088
CAS Number: 2039-86-3
EINECS: 218-025-2

Order Now

Specification Test Results

Boiling point 74-75°C/3mm
Density 1.406
ND20 1.591

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR