2-Bromostyrene ≥96% stabilized

Supplier: Matrix Scientific

006665-5G 006665-25G
101790-882EA 100 USD
101790-882 101790-884
2-Bromostyrene ≥96% stabilized
2-Bromostyrene

Hazard code(s): Irritant, Stored under Argon, Refrigerate

Formula: C₈H₇Br
MW: 183.05 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 7 °C
Density: 1.409 g/cm³ (15 °C)
MDL Number: MFCD00000076
CAS Number: 2039-88-5
EINECS: 218-027-3

Order Now

Specification Test Results

Boiling point 102-104°C/2
Density 1.416
ND20 1.5927

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR