3-(p-Tolyl)propionic acid ≥98%

Supplier: Matrix Scientific

006650-5G 006650-25G
101790-862EA 84.62 USD
101790-862 101790-864
3-(p-Tolyl)propionic acid ≥98%
3-(p-Tolyl)propionic acid

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₂
Melting Pt: 118…120 °C
MDL Number: MFCD00016561
CAS Number: 1505-50-6
EINECS: 216-132-9

Order Now

Specification Test Results

Melting point 118-120°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR