3-(m-Tolyl)propionic acid ≥98%

Supplier: Matrix Scientific

006649-1G 006649-5G
101790-858EA 75.38 USD
101790-858 101790-860
3-(m-Tolyl)propionic acid ≥98%
3-(m-Tolyl)propionic acid

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₂
Melting Pt: 41…44 °C
MDL Number: MFCD00060326
CAS Number: 3751-48-2

Order Now

Specification Test Results

Melting point 41-44°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR