3-(o-Tolyl)propionic acid ≥98%

Supplier: Matrix Scientific

006648-1G 006648-5G
101790-854EA 43.08 USD
101790-854 101790-856
3-(o-Tolyl)propionic acid ≥98%
3-(o-Tolyl)propionic acid

Hazard code(s): Irritant

Formula: C₁₀H₁₂O₂
Melting Pt: 104…106 °C
MDL Number: MFCD00079773
CAS Number: 22084-89-5

Order Now

Specification Test Results

Melting point 104-106°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR