2-Iodomesitylene ≥99%

Supplier: Matrix Scientific

006635-5G 006635-25G 006635-100G
101790-848EA 18.46 USD
101790-848 101790-850 101790-852
2-Iodomesitylene ≥99%
2-Iodomesitylene

Hazard code(s): Light Sensitive

Formula: C₉H₁₁I
Melting Pt: 27…29 °C
MDL Number: MFCD00013707
CAS Number: 4028-63-1
EINECS: 223-709-9

Order Now

Specification Test Results

Melting point 27-29°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR