Tetramethylammonium fluoride tetrahydrate ≥98% for synthesis
Supplier: Matrix Scientific
006625-25G
006625-100G
101790-832EA
112.31
USD
101790-832
101790-834
Tetramethylammonium fluoride tetrahydrate ≥98% for synthesis
Tetramethylammonium fluoride tetrahydrate
Hazard code(s): Toxic
Formula:
C₄H₁₂FN·4H₂O MW: 165.21 g/mol Melting Pt: 39…42 (42…46) °C Density: 1.115 g/cm³ (20 °C) |
MDL Number:
MFCD00149968 CAS Number: 17787-40-5 EINECS: 206-769-0 UN: 2811 ADR: 6.1 III |
Specification Test Results
Melting point | 39-42°C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...