(2-Benzyloxyphenyl)acetic acid ≥98%

Supplier: Matrix Scientific

006568-1G 006568-5G
101790-766EA 89.23 USD
101790-766 101790-768
(2-Benzyloxyphenyl)acetic acid ≥98%
(2-Benzyloxyphenyl)acetic acid

Hazard code(s): Irritant

Formula: C₆H₅CH₂OC₆H₄CH₂COOH
Melting Pt: 96…98 °C
MDL Number: MFCD00016822
CAS Number: 22047-88-7
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Melting point 96-98°C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR