2-Iodo-p-xylene tech

Supplier: Matrix Scientific
Warning

Synonyms: 2-Iodo-1,4-dimethylbenzene, 1,4-Dimethyl-2-iodobenzene

006563-5G 006563-25G
101790-758EA 176.92 USD
101790-758 101790-760
2-Iodo-p-xylene tech
2-Iodo-p-xylene

Hazard code(s): Irritant

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 230 °C
Density: 1.61 g/cm³
MDL Number: MFCD00013708
CAS Number: 1122-42-5
EINECS: 214-347-2

Order Now

Specification Test Results

Boiling point 230°C
Density 1.61

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR