4-Iodo-m-xylene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 2,4-Dimethyliodobenzene, 1-Iodo-2,4-dimethylbenzene, 2,4-Dimethyl-1-iodobenzene, 1,3-Dimethyl-4-iodobenzene

006558-5G 006558-25G 006558-100G
101790-742EA 47.69 USD
101790-742 101790-744 101790-746
4-Iodo-m-xylene ≥98%
4-Iodo-m-xylene

Hazard code(s): Irritant

Formula: C₈H₉I
MW: 232.06 g/mol
Boiling Pt: 232 °C
Density: 1.6609 g/cm³ (13 °C)
MDL Number: MFCD00013706
CAS Number: 4214-28-2
EINECS: 224-141-4

Order Now

Specification Test Results

Boiling point 232°C
Density 1.63
ND20 1.598

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR