1-Benzyloxy-4-chlorobenzene ≥98%

Supplier: Matrix Scientific

006554-5G 006554-25G
101790-730EA 52.31 USD
101790-730 101790-732
1-Benzyloxy-4-chlorobenzene ≥98%
1-Benzyloxy-4-chlorobenzene

Hazard code(s): Irritant

Formula: C₁₃H₁₁ClO
Melting Pt: 72…75 °C
MDL Number: MFCD00079702
CAS Number: 7700-27-8

Order Now

Specification Test Results

Melting point 72-75°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR