4-(Benzyloxy)benzoic acid ≥98%

Supplier: Matrix Scientific

006548-5G 006548-25G
101790-710EA 43.08 USD
101790-710 101790-712
4-(Benzyloxy)benzoic acid ≥98%
4-(Benzyloxy)benzoic acid

Hazard code(s): Irritant

Formula: C₁₄H₁₂O₃
MW: 228.25 g/mol
Melting Pt: 186…188 °C
MDL Number: MFCD00016527
CAS Number: 1486-51-7
EINECS: 216-066-0

Order Now

Specification Test Results

Melting point 186-188°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR