3'-(Trifluoromethyl)propiophenone ≥97%
Supplier: Matrix Scientific
Synonyms:
1-(3-(Trifluoromethyl)phenyl)propan-1-one, m-(Trifluoromethyl)propiophenone
006495-5G
006495-25G
101790-638EA
29.23
USD
101790-638
101790-640
3'-(Trifluoromethyl)propiophenone ≥97%
3'-(Trifluoromethyl)propiophenone
Hazard code(s): Irritant
Formula:
C₁₀H₉F₃O MW: 202.18 g/mol Boiling Pt: 72…73 °C (5mm) Melting Pt: 18…19 °C |
MDL Number:
MFCD00039227 CAS Number: 1533-03-5 |
Specification Test Results
Boiling point | 72-73°C/5mm |
Melting point | 18-19°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...