4'-(Trifluoromethyl)propiophenone ≥97%

Supplier: Matrix Scientific

Synonyms: p-(Trifluoromethyl)propiophenone, 1-(4-(Trifluoromethyl)phenyl)propan-1-one

006493-5G 006493-25G
101790-634EA 90.77 USD
101790-634 101790-636
4'-(Trifluoromethyl)propiophenone ≥97%
4'-(Trifluoromethyl)propiophenone

Hazard code(s): Irritant

Formula: C₁₀H₉F₃O
MW: 202.18 g/mol
Boiling Pt: 216 °C
Melting Pt: 36…37 °C
MDL Number: MFCD00039231
CAS Number: 711-33-1

Order Now

Specification Test Results

Boiling point 216°C
Melting point 36-37°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR