3'-(Trifluoromethyl)acetophenone ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: m-(Trifluoromethyl)acetophenone, 1-[3-(Trifluoromethyl)phenyl]ethan-1-one

006492-25G 006492-100G
101790-630EA 33.85 USD
101790-630 101790-632
3'-(Trifluoromethyl)acetophenone ≥99%
3'-(Trifluoromethyl)acetophenone

Hazard code(s): Irritant

Formula: C₉H₇F₃O
MW: 188.15 g/mol
Boiling Pt: 198…200 °C
Density: 1.23 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000391
CAS Number: 349-76-8
EINECS: 206-490-4

Order Now

Specification Test Results

Boiling point 198-200°C
Density 1.235
ND20 1.4617

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR