2'-(Trifluoromethyl)acetophenone ≥99%
Supplier: Matrix Scientific
006491-5G
006491-25G
006491-100G
101790-624EA
23.08
USD
101790-624
101790-626
101790-628
2'-(Trifluoromethyl)acetophenone ≥99%
2'-(Trifluoromethyl)acetophenone
Hazard code(s): Irritant, Refrigerate
Formula:
C₉H₇F₃O Boiling Pt: 163…164 °C Melting Pt: 15…17 °C Density: 1.255 g/cm³ |
MDL Number:
MFCD00000378 CAS Number: 17408-14-9 EINECS: 241-434-2 |
Specification Test Results
Boiling point | 163-164°C |
Density | 1.255 |
ND20 | 1.4584 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...