3'-Trifluoromethoxyacetophenone ≥98%
Supplier: Matrix Scientific
006486-1G
006486-5G
006486-25G
101790-612EA
27.69
USD
101790-612
101790-614
101790-616
3'-Trifluoromethoxyacetophenone ≥98%
3'-Trifluoromethoxyacetophenone
Hazard code(s): Irritant
Formula:
C₉H₇F₃O₂ Boiling Pt: 49…51 °C (13mm) Density: 1.285 g/cm³ |
MDL Number:
MFCD00042403 CAS Number: 170141-63-6 |
Specification Test Results
Boiling point | 49-51°C/13m |
Density | 1.285 |
ND20 | 1.452 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...